Learn More
Troemner™ Stainless Steel Electronic Balance Calibration Weights, Class 1 with Statement of Accuracy
Precision-crafted of stainless steel, with a highly polished finish
Manufacturer:  TroemnerSupplier Diversity Partner 8142
 View more versions of this product
Catalog No. 02-215-17E
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Two-Piece Electronic Balance Calibration Weight
ASTM Class 1
150g
150g
Polypropylene case
1
Statement of accuracy, ISO9001
Stainless Steel
7.95g/cm3 at 20°C
Straight cylinder
Each