Learn More
Troemner™ Stainless Steel Electronic Balance Calibration Weights, Class 1 with Traceable Certificate
Precision-crafted of stainless steel, with a highly polished finish
Manufacturer:  TroemnerSupplier Diversity Partner 8100T
 View more versions of this product
Catalog No. 02-215-7ET
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Two piece electronic balance calibration weight
ASTM Class 1
30kg
30kg
Without case
1
Traceable certificate, ISO9001
303 Stainless Steel
7.85g/cm3 at 20°C
Cylindrical with recessed grip
Each