Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troemner™ Stainless Steel Electronic Balance Calibration Weights, Class 3 with Traceable Certificate

Precision-crafted of stainless steel, with a highly polished finish

$132.00 - $2187.00

Specifications

Certifications/Compliance Traceable certificate, ISO9001
Classification ASTM Class 3
Density 7.95g/cm3 at 20°C
Product Type Two-Piece Electronic Balance Calibration Weight
Quantity Each
View More Specs

Products 7
Catalog Number Mfr. No. Includes Mass Shape Price Quantity & Availability  
Catalog Number Mfr. No. Includes Mass Shape Price Quantity & Availability  
02-216-551
View Documents
Troemner™
8300T
Without case 30 kg Cylindrical with Recessed Grip
Each for $2,187.00
Add to cart
 
02-216-553
View Documents
Troemner™
8310T
Without case 10 kg Cylindrical with Recessed Grip
Each for $1,098.00
Add to cart
 
02-216-554
View Documents
Troemner™
8334T
Polypropylene case 400 g Straight Cylinder
Each for $210.00
Add to cart
 
02-216-558
View Documents
Troemner™
8342T
Polypropylene case 150 g Straight Cylinder
Each for $185.00
Add to cart
 
02-216-562
View Documents
Troemner™
8353T
Polypropylene case 40 g Straight Cylinder
Each for $132.00
Add to cart
 
02-216-560
View Documents
Troemner™
8352T
Polypropylene case 50 g Straight Cylinder
Each for $137.00
Add to cart
 
02-216-556
View Documents
Troemner™
8340T
Polypropylene case 160 g Straight Cylinder
Each for $193.00
Add to cart
 
Description

Description

All electronic balance weights are manufactured with adjusting cavities

  • 5kg are cylindrical in shape with a groove near the top to allow for easy handling and gripping
  • Weights below 1kg are straight cylinders
  • Fractional weights 500 mg to 5 mg are made of stainless steel (30 mg weights are aluminum); fractional weights 3 mg and below are made of aluminum
  • Weights 5kg and below supplied in a rugged, precision-fit, high quality polycarbonate case with handling glove
  • With Traceable Certificate

Specifications

Specifications

Traceable certificate, ISO9001
7.95g/cm3 at 20°C
Each
ASTM Class 3
Two-Piece Electronic Balance Calibration Weight
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit