Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troemner™ Stainless Steel Electronic Balance Calibration Weights Sets

100g to 1kg

Manufacturer:  Troemner™ 8170

 View more versions of this product

Catalog No. 02-215-8


  Check Availability
Add to cart

Description

Description

  • 2 piece constructions
  • Weights below 1 kg are straight cylinders
  • 1kg weights have groove neare thet top for easy handling
  • Weight set configurations: 1kg, 500g, 300g, 200g, 100g
  • Total capacity of weight set: 2100g

  • UltraClass: With weight tolerances 50% greater than ANSI/ASTM E617 Class 1 tolerances
  • ANSI/ASTM Class 1: Can be used as a reference standard in calibrating other weights and is appropriate for calibrating high precision analytical balances with a readability as low as 0.1mg to 0.01mg
  • ANSI/ASTM Class 2: Appropriate for calibrating high-precision top loading balances with a readability as low as 0.01g to 0.001g
  • ANSI/ASTM Class 3: Appropriate for calibrating balances with moderate precision, with a readability as low as 0.1g to 0.01g
  • ANSI/ASTM Class 4: For calibration of semi-analytical balances and for student use
    • Order Info

      Supplied with impact-resistant polypropylene case.

Specifications

Specifications

Electronic Balance Calibration Weight
Class 1
100g to 1kg
100g to 1kg
5/Set
Statement of accuracy, ISO9001
Stainless Steel
7.95g/cm3 at 20°C
Cylindrical weight: 1kg, 500g, 300g, 200g, 100g, Plastic case, white Cotton glove
Multiple
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit