Learn More
Troemner™ Stainless Steel Electronic Balance Calibration Weights, UltraClass with Traceable Certificate
Precision-crafted of stainless steel, with a highly polished finish
$149.10 - $326.50
Specifications
Certifications/Compliance Traceable certificate, ISO9001
Classification UltraClass
Quantity Each
Shape Cylindrical with Recessed Grip
View More Specs
Products 3
Catalog Number Mfr. No. Product Type Includes Mass Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Includes Mass Price Quantity & Availability  
02-214-7H
View Documents
Troemner™
8028T
Two-Piece Electronic Balance Calibration Weight Polypropylene case 1000 g Each for $326.50
Add to cart
02-214-7P
View Documents
Troemner™
8044T
Two-Piece Electronic Balance Calibration Weight Polypropylene case 100 g Each for $168.10
Add to cart
02-214-7Q
View Documents
Troemner™
8052T
Two-Piece Electronic Balance Calibration Weight Polypropylene case 50 g Each for $149.10
Add to cart
Description
Description
UltraClass weights are available with weight tolerances 50% greater than ANSI/ASTM E617 Class 1 tolerances. They combine high precision with the advantage of two-piece construction (1g and larger), avoiding costly replacement issues associated with one-piece weights.
  • 5kg are cylindrical in shape with a groove near the top to allow for easy handling and gripping
  • Weights below 1kg are straight cylinders
  • Weights 5kg and below supplied in a rugged, precision-fit, high quality polycarbonate case with handling glove
  • With Traceable Certificate
Specifications
Specifications
Traceable certificate, ISO9001
UltraClass
Each
Cylindrical with Recessed Grip