Learn More
Troemner™ Talboys™ 134 Series Heavy Duty Laboratory Mixers Non-distribution product as customer accommodation.
With triple shaft design
Manufacturer:  Troemner™ 30531169
Catalog No. 22-267344
Add to cart
Description
Description
  • For mixing high viscous materials in small quantities
  • Offers high torque for slow speed mixing
  • Maintains constant speed during viscosity changes
  • Comes with adjustable Jacobs style chuck that accepts accessory shaft
  • Available with mounting rod with wrench and carriage bolt for mounting
Specifications
Specifications
500 to 7500rpm
Mixing high viscous materials in small quantities
0.79cm
0.5
120V
1/18
0.31 in.