Learn More
Troemner™ Talboys™ Heavy-Duty Lab-Lifts
With over-sized, sure-grip adjustment knobs that provide extra leverage for easy height adjustments
Manufacturer:  Troemner™ 960054
 View more versions of this product
Catalog No. 02-214-201
  Check Availability
Add to cart
Description
Description
  • Heavy duty 300 series stainless steel construction
  • Chemical resistant
  • Autoclavable
  • Hold variety of items such as glassware, hotplates, baths and magnetic stirrers
  • Suited for use in fume hoods or bench tops
  • Can withstand extreme environments and high load applications
  • Available in different sizes
Specifications
Specifications
Yes
5 to 2.5 in.
Stainless Steel
3 × 3 in. (7.62 × 7.62cm)
20 lb.
7.62 to 7.62cm
Talboys™ Heavy Duty Lab Lifts