Learn More
Troemner™ Talboys™ Labjaws™ Bench Clamps Available on GSA/VA Contract for Federal Government customers only.
Durable, cast-aluminum clamps secure gas cylinders to benches, tables, or other flat surfaces
Manufacturer:  Troemner™ 970035
 View more versions of this product
Catalog No. 02-216-359
Add to cart
Description
Description
  • Includes a one-inch (2.5cm) wide, 54 in. (137.2cm) long nylon strap with a nickel plated, nonslip spring catch and buckle for simple adjustment around 4 to 14 in. (10.2 to 35.6cm) diameter cylinders
Model 711 Bench Clamps
  • Attach to surfaces up to 2.5 in. (6.4cm) thick
  • Offer an easy to grip tightening handle and a nylon pad to prevent surface damage
  • Furnished with two chamfered mounting screw holes for permanent surface attachement
  • Available with “Secure Cylinder” safety reminder
Model 716 Bench Clamps
  • Offers the same features as Model 711, plus a durable 49 in. (124.5cm) safety chain for additional security
  • Model 712 Heavy-Duty Bench Clamp
    • Strengthened by two screw clamps that tighten for extra firm mounting to flat surfaces up 1.75 in. (4.4cm) thick with a 1.25 in. (3.2cm) overhang
    • Ideal for temporary storage locations
    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
    Specifications
    Specifications
    54 L x 1 in. W Nylon strap with a Nickel plated, non-slip spring catch and buckle for simple adjustment around cylinders, strap
    8.25cm
    Heavy Duty Bench Clamp w/Strap
    3.25 in.
    Cast Aluminum