Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troemner™ Troemner™ Talboys™ Labjaws™ Stainless Steel Double Buret Clamp

Stainless steel electro-polished finish clamp holds any size buret from micro to 100mL capacity

Manufacturer:  Troemner™ 916367

 View more versions of this product

Catalog No. 02-217-129


Add to cart

Description

Description

  • Holds any size buret from micro to 100mL
  • Simply compress scissor-like mechanism, insert buret and gently release to grip
  • Numbers and graduation on buret remain easy to read
  • For height adjustments, recompress mechanism and slide buret up or down and gently release
  • Clamp unit attaches to optional standard support rod with built-in reinforced hook connector
  • Adjusting nut faces forward for easy use
  • Stainless steel support rod attaches to optional porcelain base
  • Support rod: 0.51 dia. x 22.76 in.L (13 x 578mm)
  • Porcelain base: 7L x 13W x 1 in.H (178 x 330 x 25mm)

Specifications

Specifications

Double Buret Clamp
Stainless Steel
Each
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit