Learn More
Troemner™ Three-Prong Dual Adjust Swivel Clamp Available on GSA/VA Contract for Federal Government customers only.
Swivel clamps have an integral holder for attaching to a lab-frame or other apparatus.
Manufacturer:  Troemner™ 916066
 View more versions of this product
Catalog No. 02-300-209
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Vinyl and Fiberglass sleeves prong covers
178mm
Swivel Clamp
0 to 2.72 in.
7.01 in.
Nickel-plated zinc
Medium
0 to 69mm