Learn More
Troemner™ UltraClass Gold Individual Analytical Precision Weights with NVLAP Certificate
For calibrating high-precision analytical balances with readability as low as .001mg
Manufacturer:  Troemner™ 7035UCGW
 View more versions of this product
Catalog No. 02-214-246
Add to cart
Description
Description
Made of Troemner Alloy 8, a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage. Exclusive material has a density of 8.0 at 25°C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to -195.5°C (-320°F).
  • Weight tolerance is equal to ANSI/ASTM E617 Class 00 and meets or exceeds OIML R 111 Class E1
  • This class is used as a reference standard for calibrating other reference standards and weights where the stability of the environment and careful handling are assured
  • Appropriate for calibrating high-precision analytical balances with a readability as low as .001mg
  • Weights come with a Positive Calibration Tolerance Guarantee
Note: Fractional weights 5 to 500mg are made of stainless steel (except 30mg, which are aluminum). Weights 3mg and below are made of aluminum.
Certifications
With NVLAP™ weight calibration certificate.
Specifications
Specifications
Metric analytical sheet weight with one side turned up
UltraClass Gold
3mg
3mg
Polycarbonate case
NVLAP, ISO9001
Aluminum
2.7g/cm3 at 20°C
Flat with one end turned up for easy handling
Each