Learn More
Troemner™ UltraClass Gold Individual Analytical Precision Weights with NVLAP Certificate
For calibrating high-precision analytical balances with readability as low as .001mg
Specifications
Certifications/Compliance NVLAP, ISO9001
Classification UltraClass Gold
Includes Polycarbonate case
Quantity Each
View More Specs
Products 5
Catalog Number Mfr. No. Material Mass Shape Price Quantity & Availability  
Catalog Number Mfr. No. Material Mass Shape Price Quantity & Availability  
02-214-258
View Documents
Troemner™
7016UCGW
Stainless Steel 200 g Cylindrical with Handling Knob This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
02-214-264
View Documents
Troemner™
7020UCGW
Stainless Steel 20 g Cylindrical with Handling Knob This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
02-214-270
View Documents
Troemner™
7012UCGW
Stainless Steel 2 kg Cylindrical with Handling Knob This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
02-214-243
View Documents
Troemner™
7019UCGW
Stainless Steel 30 g Cylindrical with Handling Knob This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
02-214-216
View Documents
Troemner™
7014UCGW
Stainless Steel 500 g Cylindrical with Handling Knob This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Made of Troemner Alloy 8, a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage. Exclusive material has a density of 8.0 at 25°C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to -195.5°C (-320°F).
  • Weight tolerance is equal to ANSI/ASTM E617 Class 00 and meets or exceeds OIML R 111 Class E1
  • This class is used as a reference standard for calibrating other reference standards and weights where the stability of the environment and careful handling are assured
  • Appropriate for calibrating high-precision analytical balances with a readability as low as .001mg
  • Weights come with a Positive Calibration Tolerance Guarantee
Note: Fractional weights 5 to 500mg are made of stainless steel (except 30mg, which are aluminum). Weights 3mg and below are made of aluminum.
Certifications
With NVLAP™ weight calibration certificate.
Specifications
Specifications
NVLAP, ISO9001
UltraClass Gold
Polycarbonate case
Each