Learn More
Troemner™ UltraClass Platinum Calibration Weight Sets
For calibrating high-precision analytical balances with readability as low as 0.0001mg
Manufacturer:  Troemner™ 7236UCP
 View more versions of this product
Catalog No. 02-214-510
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Made of Troemner Alloy 8, a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage. Exclusive material has a density of 8.0 at 25°C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to -195.5°C (-320°F).
  • Weight tolerance is equal to ANSI/ASTM E617 Class 000 and Class E0
  • This class is used as a reference standard for calibrating other reference standards and weights where the stability of the environment and careful handling are assured
  • Appropriate for calibrating high-precision analytical balances with a readability as low as 0.0001mg
  • Weights come with a Positive Calibration Tolerance Guarantee
Construction:
  • Fractional weights 5 to 500mg are made of stainless steel (except 30mg, which are aluminum)
  • Weights 3mg and below are made of aluminum
Note: Weight sets where the largest weight is 500g and below are supplied in a high-quality polycarbonate case. A wood case is provided for sets where the largest weight is 1kg and above.
Certifications
Includes Statement of Accuracy.
Specifications
Specifications
Medium metric analytical weight set
UltraClass Platinum
10mg to 20g
Cylindrical weight: 20g, 10g, 5g, 2 x 2g, 1g, sheet weight: 500mg, 2 x 200mg, 100mg, 50mg, 2 x 20mg, 10mg, Polycarbonate case
Statement of accuracy, ISO9001
Stainless Steel
20g-10mg
14/Set