Troemner™ UltraClass Platinum Electronic Balance Calibration Weights

Constructed of special Troemner Alloy 8 stainless steel

Manufacturer: TroemnerSupplier Diversity Partner 8028UCP

 View more versions of this product

Catalog No. 02-218-125

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    Product Type Two piece electronic balance calibration weight
    Certifications/Compliance Statement of accuracy, ISO9001
    Includes Polypropylene case
    Classification UltraClass Platinum
    Quantity Each
    Material 316 Stainless Steel
    Weight (Metric) 1kg
    Density 7.95g/cm3 at 20°C
    Mass 1kg
    Shape Cylindrical with groove handle

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit