Learn More
Troemner™ UltraClass Platinum Electronic Balance Calibration Weights with Traceable Certificate
Constructed of special Troemner Alloy 8 stainless steel
Manufacturer:  TroemnerSupplier Diversity Partner 8024UCPT
 View more versions of this product
Catalog No. 02-218-127
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
Two-Piece Electronic Balance Calibration Weight
UltraClass Platinum
2kg
2kg
Polypropylene case
Traceable certificate, ISO9001
Stainless Steel
7.85g/cm3 at 20°C
Cylindrical with groove handle
Each