Learn More
Troemner™ UltraClass Platinum Individual Analytical Precision Weights
For calibrating high-precision analytical balances with readability as low as 0.0001mg
Specifications
Certifications/Compliance Statement of accuracy, ISO9001
Classification UltraClass Platinum
Includes Polycarbonate case
Quantity Each
View More Specs
Products 3
Catalog Number Mfr. No. Product Type Mass Shape Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Mass Shape Price Quantity & Availability  
02-214-439
View Documents
Troemner™
7033UCP
Metric Analytical Sheet Weight with One Side Turned Up 10 mg Flat with one end turned up for easy handling This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
02-214-433
View Documents
Troemner™
7029UCP
Metric Analytical Sheet Weight with One Side Turned Up 100 mg Flat with one end turned up for easy handling This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
02-214-451
View Documents
Troemner™
7025UCP
Metric Analytical Weight with Handling Knob 1 g Cylindrical with Handling Knob This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
Description
Description
Made of Troemner Alloy 8, a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage. Exclusive material has a density of 8.0 at 25°C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to -195.5°C (-320°F).
  • Weight tolerance is equal to ANSI/ASTM E617 Class 000 and Class E0
  • This class is used as a reference standard for calibrating other reference standards and weights where the stability of the environment and careful handling are assured
  • Appropriate for calibrating high-precision analytical balances with a readability as low as 0.0001mg
  • Weights come with a Positive Calibration Tolerance Guarantee
Note: Fractional weights 5 to 500mg are made of stainless steel (except 30mg, which are aluminum). Weights 3mg and below are made of aluminum.
Certifications
Includes Statement of Accuracy.
Specifications
Specifications
Statement of accuracy, ISO9001
UltraClass Platinum
Polycarbonate case
Each