Troemner™ UltraClass Platinum Individual Analytical Precision Weights with NVLAP Certificate
For calibrating high-precision analytical balances with readability as low as 0.0001mg
Manufacturer: TroemnerSupplier Diversity Partner 7020-UCPW
 View all options for this product
Catalog No. 02-214-420
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to for assistance. 
    Product Type Metric analytical weight with handling knob
    Certifications/Compliance NVLAP, ISO9001
    Material Alloy 8 Stainless Steel
    Weight (Metric) 20g
    Density 8.03g/cm3 at 20°C
    Mass 20g
    Shape Cylindrical with handling knob
    Includes Polycarbonate case
    Quantity Each
    Made of Troemner Alloy 8, a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage. Exclusive material has a density of 8.0 at 25°C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to -195.5°C (-320°F).
    • Weight tolerance is equal to ANSI/ASTM E617 Class 000 and Class E0
    • This class is used as a reference standard for calibrating other reference standards and weights where the stability of the environment and careful handling are assured
    • Appropriate for calibrating high-precision analytical balances with a readability as low as 0.0001mg
    • Weights come with a Positive Calibration Tolerance Guarantee
    Note: Fractional weights 5 to 500mg are made of stainless steel (except 30mg, which are aluminum). Weights 3mg and below are made of aluminum.
  • Description & Specifications
Related Products
Product Content Feedback