Learn More
Troemner™ UltraClass Platinum Individual Analytical Precision Weights with Traceable Certificate
For calibrating high-precision analytical balances with readability as low as 0.0001mg
Manufacturer:  Troemner™ 7012UCPT
 View more versions of this product
Catalog No. 02-214-428
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Made of Troemner Alloy 8, a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage. Exclusive material has a density of 8.0 at 25°C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to -195.5°C (-320°F).
  • Weight tolerance is equal to ANSI/ASTM E617 Class 000 and Class E0
  • This class is used as a reference standard for calibrating other reference standards and weights where the stability of the environment and careful handling are assured
  • Appropriate for calibrating high-precision analytical balances with a readability as low as 0.0001mg
  • Weights come with a Positive Calibration Tolerance Guarantee
Note: Fractional weights 5 to 500mg are made of stainless steel (except 30mg, which are aluminum). Weights 3mg and below are made of aluminum.
Certifications
With Traceable certificate.
Specifications
Specifications
Metric analytical weight with handling knob
UltraClass Platinum
2kg
2kg
Polycarbonate case
Traceable certificate, ISO9001
Stainless Steel
8.03g/cm3 at 20°C
Cylindrical with handling knob
Each