Learn More
Troemner™ UltraClass Platinum Individual Analytical Precision Weights with Traceable Certificate
For calibrating high-precision analytical balances with readability as low as 0.0001mg
Manufacturer:  TroemnerSupplier Diversity Partner 7033UCPT
 View more versions of this product
Catalog No. 02-214-440
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
Metric analytical sheet weight with one side turned up
UltraClass Platinum
10mg
10mg
Polycarbonate case
Traceable certificate, ISO9001
Stainless Steel
7.95g/cm3 at 20°C
Flat with one end turned up for easy handling
Each