Learn More
Troemner™ UltraClass Platinum Individual Analytical Precision Weights with Traceable Certificate
For calibrating high-precision analytical balances with readability as low as 0.0001mg
Specifications
Certifications/Compliance Traceable certificate, ISO9001
Classification UltraClass Platinum
Includes Polycarbonate case
Quantity Each
View More Specs
Products 1
Catalog Number Mfr. No. Product Type Mass Shape Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Mass Shape Price Quantity & Availability  
02-214-416
View Documents
Troemner™
7016UCPT
Metric Analytical Weight with Handling Knob 200 g Cylindrical with Handling Knob This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
Description
Description
Made of Troemner Alloy 8, a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage. Exclusive material has a density of 8.0 at 25°C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to -195.5°C (-320°F).
  • Weight tolerance is equal to ANSI/ASTM E617 Class 000 and Class E0
  • This class is used as a reference standard for calibrating other reference standards and weights where the stability of the environment and careful handling are assured
  • Appropriate for calibrating high-precision analytical balances with a readability as low as 0.0001mg
  • Weights come with a Positive Calibration Tolerance Guarantee
Note: Fractional weights 5 to 500mg are made of stainless steel (except 30mg, which are aluminum). Weights 3mg and below are made of aluminum.
Certifications
With Traceable certificate.
Specifications
Specifications
Traceable certificate, ISO9001
UltraClass Platinum
Polycarbonate case
Each