Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trolamine, NF, 99-107.4%, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
102-71-6
C6H15NO3
149.19
GSEJCLTVZPLZKY-UHFFFAOYSA-N
2-[bis(2-hydroxyethyl)amino]ethan-1-ol
OCCN(CCO)CCO
102-71-6
C6H15NO3
149.19
GSEJCLTVZPLZKY-UHFFFAOYSA-N
2-[bis(2-hydroxyethyl)amino]ethan-1-ol
OCCN(CCO)CCO

TR143, 102-71-6, C6H15NO3

Spectrum™ Trolamine, NF is used as an emulsifying excipient in hand and body lotions, shaving creams, soaps, and shampoos. All Spectrum Chemical NF products, including all NF Trolamine products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Poly Drum
Water 0.005
Ignition Residue 0.0005
Refractive Index 1.481-1.486
Quantity 20 L
CAS Max % 1
Percent Purity 99 to 107.4%
Grade NF