Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tromethamine, Multi-Compendial, U.S.P., J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
77-86-1
C4H11NO3
121.136
LENZDBCJOHFCAS-UHFFFAOYSA-N
trometamol,tris,tromethamine,tris hydroxymethyl aminomethane,tham,trisamine,tris buffer,trizma,2-amino-2-hydroxymethyl propane-1,3-diol,tris base
6503
CHEBI:9754
2-amino-2-(hydroxymethyl)propane-1,3-diol
C(C(CO)(CO)N)O
77-86-1
C4H11NO3
121.136
LENZDBCJOHFCAS-UHFFFAOYSA-N
trometamol,tris,tromethamine,tris hydroxymethyl aminomethane,tham,trisamine,tris buffer,trizma,2-amino-2-hydroxymethyl propane-1,3-diol,tris base
6503
CHEBI:9754
2-amino-2-(hydroxymethyl)propane-1,3-diol
C(C(CO)(CO)N)O
Quantity 500g
Packaging Poly Bottle
Formula Weight 121.14
Grade Multi-Compendial/USP
Chemical Name or Material Tromethamine