Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TROP-2 Alexa Fluor 405-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
Supplier: R&D Systems FAB650V100UG
This item is not returnable.
View return policy
Description
TROP-2 Monoclonal specifically detects TROP-2 in Human samples. It is validated for Flow Cytometry.Specifications
TROP-2 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P09758 | |
TACSTD2 | |
Mouse myeloma cell line NS0-derived recombinant human TROP-2 His27-Thr274 Accession # P09758 | |
RUO | |
4070 | |
Human | |
IgG2a |
Flow Cytometry | |
77220 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Cell surface glycoprotein Trop-2, EGP1, EGP-1, epithelial glycoprotein-1, GA733-1, GA733-1GP50, gastrointestinal tumor-associated antigen GA7331, gp50, M1S1cell surface glycoprotein TROP2, Membrane component chromosome 1 surface marker 1, membrane component, chromosome 1, surface marker 1, Pancreatic carcinoma marker protein GA733-1, pancreatic carcinoma marker protein GA7331,40kD glycoprotein, identified by monoclonal GA733, T16, TACSTD2, TROP2, TROP2GA7331, tumor-associated calcium signal transducer 2 | |
Mouse | |
100 μg | |
Primary | |
Detects human TROP-2 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) VCAM-1 or rhICAM-1 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction