Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tropanyl-3,5-dimethylbenzoate, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
100mg
This item is not returnable. View return policy
85181-40-4
C17H23NO2
273.376
HDDNYFLPWFSBLN-XYPWUTKMSA-N
tropanserin,tropyl 3,5-dimethylbenzoate,tropanserina,tropanserine,tropanserinum,tropanserine french,tropanserinum latin,tropanyl 3,5-dimethylbenzoate,tropanserina spanish,tropanserin inn:ban
68600
[(1R,5S)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 3,5-dimethylbenzoate
CC1=CC(=CC(=C1)C(=O)OC2CC3CCC(C2)N3C)C
This item is not returnable. View return policy
85181-40-4
C17H23NO2
273.376
HDDNYFLPWFSBLN-XYPWUTKMSA-N
tropanserin,tropyl 3,5-dimethylbenzoate,tropanserina,tropanserine,tropanserinum,tropanserine french,tropanserinum latin,tropanyl 3,5-dimethylbenzoate,tropanserina spanish,tropanserin inn:ban
68600
[(1R,5S)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 3,5-dimethylbenzoate
CC1=CC(=CC(=C1)C(=O)OC2CC3CCC(C2)N3C)C

5-HT3 antagonist

5-HT3 antagonist.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 100mg
Formula Weight 273.37
Percent Purity >99%
Chemical Name or Material Tropanyl-3, 5-dimethylbenzoate

Recommended Storage : Store at Room Temperature