Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tropine, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
120-29-6
C8H15NO
141.21
MFCD00005551
CYHOMWAPJJPNMW-RNFRBKRXSA-N
tropine,1r,5r-8-methyl-8-azabicyclo 3.2.1 octan-3-ol,endo-8-methyl-8-azabicyclo 3.2.1 octan-3-ol,spectrum3_001840,1r,5r-8-methyl-8-azabicyclo 3.2.1 octan-3beta-ol
6101956
(1R,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-ol
CN1C2CCC1CC(C2)O
120-29-6
C8H15NO
141.21
MFCD00005551
CYHOMWAPJJPNMW-RNFRBKRXSA-N
tropine,1r,5r-8-methyl-8-azabicyclo 3.2.1 octan-3-ol,endo-8-methyl-8-azabicyclo 3.2.1 octan-3-ol,spectrum3_001840,1r,5r-8-methyl-8-azabicyclo 3.2.1 octan-3beta-ol
6101956
(1R,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-ol
CN1C2CCC1CC(C2)O
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25g
Water 2% max. (K.F.)
Density 1.0400g/mL
Color White to Yellow
Specific Gravity 1.04
Boiling Point 233.0°C
Melting Point 62.0°C to 64.0°C
Assay Percent Range 98%
Beilstein 21, 16
Merck Index 15, 9960
Solubility Information Solubility in water: 100g/L (20°C). Other solubilities: soluble in ether and chloroform, freely soluble in alcohol
Formula Weight 141.21
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material Tropine
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use only outdoors or in a well-ventilated area.
Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning