Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tropinone 99%, ACROS Organics™

Click to view available options
Quantity:
5g
10g
50g
Packaging:
Glass bottle
532-24-1
C8H13NO
139.2
MFCD00005549
QQXLDOJGLXJCSE-UHFFFAOYSA-N
tropinone,8-methyl-8-azabicyclo 3.2.1 octan-3-one,3-tropanone,tropanone,tropanon,tropinon,tropionone,3-tropinone,8-azabicyclo 3.2.1 octan-3-one, 8-methyl,1.alpha.h,5.alpha.h-tropan-3-one
79038
8-methyl-8-azabicyclo[3.2.1]octan-3-one
CN1C2CCC1CC(=O)C2
532-24-1
C8H13NO
139.2
MFCD00005549
QQXLDOJGLXJCSE-UHFFFAOYSA-N
tropinone,8-methyl-8-azabicyclo 3.2.1 octan-3-one,3-tropanone,tropanone,tropanon,tropinon,tropionone,3-tropinone,8-azabicyclo 3.2.1 octan-3-one, 8-methyl,1.alpha.h,5.alpha.h-tropan-3-one
79038
8-methyl-8-azabicyclo[3.2.1]octan-3-one
CN1C2CCC1CC(=O)C2
Flash Point 90°C
Infrared Spectrum Authentic
Melting Point 41.0°C to 44.0°C
Boiling Point 113.0°C (25.0 mmHg)
Quantity 10g
Color Yellow to Brown
Packaging Glass bottle
Assay Percent Range 99%
Beilstein 21,258
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol, ether, acetone, benzene, slightly soluble in chloroform
Formula Weight 139.2
Physical Form Crystals or Crystalline Powder
Percent Purity 99%
Chemical Name or Material Tropinone, 99%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 208-530-6

TSCA : TSCA