Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tropolone, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
533-75-5
C7H6O2
122.12
MFCD00004158
MDYOLVRUBBJPFM-UHFFFAOYSA-N
tropolone,purpurocatechol,2-hydroxycyclohepta-2,4,6-trienone,2,4,6-cycloheptatrien-1-one, 2-hydroxy,2-hydroxytropone,2-hydroxy-2,4,6-cycloheptatrien-1-one,2-hydroxy-2,4,6-cycloheptatrienone,unii-7l6dl16p1t,ccris 6609,tropomyosins
10789
CHEBI:79966
2-hydroxycyclohepta-2,4,6-trien-1-one
C1=CC=C(C(=O)C=C1)O
533-75-5
C7H6O2
122.12
MFCD00004158
MDYOLVRUBBJPFM-UHFFFAOYSA-N
tropolone,purpurocatechol,2-hydroxycyclohepta-2,4,6-trienone,2,4,6-cycloheptatrien-1-one, 2-hydroxy,2-hydroxytropone,2-hydroxy-2,4,6-cycloheptatrien-1-one,2-hydroxy-2,4,6-cycloheptatrienone,unii-7l6dl16p1t,ccris 6609,tropomyosins
10789
CHEBI:79966
2-hydroxycyclohepta-2,4,6-trien-1-one
C1=CC=C(C(=O)C=C1)O
Quantity 5g
Flash Point >110°C
Packaging Glass bottle
Boiling Point 115.0°C (12.0 mmHg)
Infrared Spectrum Authentic
Melting Point 50.0°C to 54.0°C
Color Beige-Cream to White
Assay Percent Range 98%
Merck Index 15, 9963
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in ethyl ether, acetone, organic solvents
Formula Weight 122.12
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material Tropolone
Show More Show Less