Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tropomyosin-1 Sheep anti-Mouse, Rat, Polyclonal, Novus Biologicals™
Sheep Polyclonal Antibody
$177.15 - $666.00
Specifications
Antigen | Tropomyosin-1 |
---|---|
Dilution | Western Blot 1:200-1:1000, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:100-1:500 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Tropomyosin-1 Polyclonal specifically detects Tropomyosin-1 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Tropomyosin-1 | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Mouse, Rat | |
P09493 | |
7168 | |
Synthetic peptide corresponding to the 9d exon from the alpha gene of tropomyosin. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot 1:200-1:1000, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:100-1:500 | |
Polyclonal | |
Sheep | |
Cytoskeleton Markers | |
PBS with No Preservative | |
alpha tropomyosin, alpha-tropomyosin, C15orf13, cardiomyopathy, hypertrophic 3, chromosome 15 open reading frame 13, CMD1Y, CMH3, HTM-alpha, sarcomeric tropomyosin kappa, TMSA, tropomyosin 1 (alpha), tropomyosin 1 (alpha) isoform 1, tropomyosin 1 (alpha) isoform 2, tropomyosin 1 (alpha) isoform 3, tropomyosin 1 (alpha) isoform 4, tropomyosin 1 (alpha) isoform 5, tropomyosin 1 (alpha) isoform 6, tropomyosin 1 (alpha) isoform 7, tropomyosin alpha-1 chain, Tropomyosin-1 | |
TPM1 | |
IgG | |
Affinity Purified | |
Recognizes Tropomyosin 1, 2, 3, 5a, 5b, 6. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title