Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tropomyosin 3 alpha chain Mouse anti-Human, Mouse, Rat, Clone: 4D10A5, MilliporeSigma™

Mouse Monoclonal Antibody

$373.00

Specifications

Antigen Tropomyosin 3 alpha chain
Clone 4D10A5
Concentration 0.5 mg/mL
Applications Western Blot
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABT1336100
View Documents
MilliporeSigma
MABT1336100UL
100 μL
Each for $373.00
Add to cart
 
MABT133625
View Documents
MilliporeSigma
MABT133625UL
25 μL
Each for $373.00
Add to cart
 
Description

Description

Tropomyosin 3 alpha chain Monoclonal antibody specifically detects Tropomyosin 3 alpha chain in Human, Mouse, Rat samples. It is validated for Western Blotting.
Specifications

Specifications

Tropomyosin 3 alpha chain
0.5 mg/mL
Unconjugated
Mouse
Human, Mouse, Rat
Diphtheria Toxoid Carrier Protein conjugated linear peptide corresponding to 27 amino acids from the C-terminal region of isoform 2 of rat Tropomyosin alpha-3 chain (Tpm 3).
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
4D10A5
Western Blot
Purified
RUO
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Monoclonal
Protein G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit