Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tropomyosin alpha-1 Mouse anti-Rat, Clone: 2F4E11, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABT1337100UG
Description
Tropomyosin alpha-1 Monoclonal antibody specifically detects Tropomyosin alpha-1 in Rat samples. It is validated for Western Blotting.Specifications
Tropomyosin alpha-1 | |
Monoclonal | |
1 mg/mL | |
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide | |
KLH-conjugated linear peptide corresponding to 19 amino acids from the C-terminal region of rat Tropomyosin alpha-1 chain, isoform 4. | |
100 μg | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
Western Blot | |
2F4E11 | |
Unconjugated | |
Mouse | |
Protein G purified | |
RUO | |
Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction