Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tropomyosin alpha-1 Mouse anti-Rat, Clone: 2F4E11, MilliporeSigma™
Mouse Monoclonal Antibody
$185.00 - $467.00
Specifications
Antigen | Tropomyosin alpha-1 |
---|---|
Clone | 2F4E11 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABT133725
|
MilliporeSigma
MABT133725UG |
25 μg |
Each for $185.00
|
|
|||||
MABT1337100
|
MilliporeSigma
MABT1337100UG |
100 μg |
Each for $467.00
|
|
|||||
Description
Tropomyosin alpha-1 Monoclonal antibody specifically detects Tropomyosin alpha-1 in Rat samples. It is validated for Western Blotting.Specifications
Tropomyosin alpha-1 | |
1 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
2F4E11 | |
Western Blot | |
Unconjugated | |
Mouse | |
Rat | |
KLH-conjugated linear peptide corresponding to 19 amino acids from the C-terminal region of rat Tropomyosin alpha-1 chain, isoform 4. | |
Protein G purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title