Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troponin I Type 1 (slow skeletal) Mouse anti-Human, Alexa Fluor™ 750, Clone: 360000000, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP222621AF750

Catalog No. NP222621A70


Add to cart

Description

Description

Troponin I Type 1 (slow skeletal) Monoclonal antibody specifically detects Troponin I Type 1 (slow skeletal) in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Troponin I Type 1 (slow skeletal)
360000000
Western Blot, ELISA
Conjugated, Purified
TNNI1
Recombinant human TNNI1 (1-187aa) purified from E. coli
Protein A purified
Cytoskeleton Markers, Stem Cell Markers
Primary
7135.0
ELISA, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
DKFZp451O223, slow-twitch isoform, SSTNI, TNN1, troponin I type 1 (skeletal, slow), troponin I, skeletal, slow, troponin I, slow skeletal muscle
Mouse
IgG2b Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit