Learn More
Troponin I Type 3 (cardiac) Mouse anti-Human, Mouse, Alexa Fluor™ 532, Clone: 2C9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP235336AF532
Catalog No. NP23336A32
Add to cart
Description
Description
Troponin I Type 3 (cardiac) Monoclonal antibody specifically detects Troponin I Type 3 (cardiac) in Human,Mouse samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
Troponin I Type 3 (cardiac)
2C9
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Cardiac troponin I, CMD1FF, CMD2A, CMH7MGC116817, cTnI, familial hypertrophic cardiomyopathy 7, RCM1, TNNC1troponin I, cardiac, troponin I type 3 (cardiac), troponin I, cardiac muscle
Mouse
IgG2b Kappa
Store at 4°C in the dark.
Monoclonal
Human, Mouse
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
TNNI3
Partial recominant human Troponin 1 (between amino acids 1-200) [UniProt P19429]
0.1 mL
Primary
7137