Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Troponin I Type 3 (cardiac) Mouse anti-Human, Mouse, Alexa Fluor™ 594, Clone: 10000000000, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP235337AF594
Description
Troponin I Type 3 (cardiac) Monoclonal specifically detects Troponin I Type 3 (cardiac) in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Troponin I Type 3 (cardiac) | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
TNNI3 | |
Partial recominant human Troponin 1 (between amino acids 1-200) [UniProt P19429] | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG2b κ |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
10000000000 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Cardiac troponin I, CMD1FF, CMD2A, CMH7MGC116817, cTnI, familial hypertrophic cardiomyopathy 7, RCM1, TNNC1troponin I, cardiac, troponin I type 3 (cardiac), troponin I, cardiac muscle | |
Mouse | |
Protein G purified | |
Apoptosis, Chemokines and Cytokines, Cytokine Research, Cytoskeleton Markers, GPCR, Immunology, Innate Immunity, Phospho Specific, Signal Transduction, Stem Cell Markers | |
7137.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title