Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troponin I Type 3 (cardiac) Mouse anti-Human, Mouse, Alexa Fluor™ 750, Clone: 2C9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP235336AF750

Catalog No. NP23336A70


Add to cart

Description

Description

Troponin I Type 3 (cardiac) Monoclonal antibody specifically detects Troponin I Type 3 (cardiac) in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

Troponin I Type 3 (cardiac)
2C9
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated, Purified
TNNI3
Partial recominant human Troponin 1 (between amino acids 1-200) [UniProt P19429]
Protein G purified
Apoptosis, Chemokines and Cytokines, Cytokine Research, Cytoskeleton Markers, GPCR, Immunology, Innate Immunity, Phospho Specific, Signal Transduction, Stem Cell Markers
Primary
7137.0
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
Cardiac troponin I, CMD1FF, CMD2A, CMH7MGC116817, cTnI, familial hypertrophic cardiomyopathy 7, RCM1, TNNC1troponin I, cardiac, troponin I type 3 (cardiac), troponin I, cardiac muscle
Mouse
IgG2b Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit