Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Troponin T, Cardiac Isoform Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Troponin T, Cardiac Isoform Ab-1, Mouse Monoclonal Antibody.

$425.50 - $902.00

Specifications

Antigen Troponin T, Cardiac Isoform Ab-1
Clone 13-11
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 7
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS295P
View Documents
Thermo Scientific™
MS295P
Purified Ab with BSA and Azide 1mL
Each for $902.00
Add to cart
 
MS295P1
View Documents
Thermo Scientific™
MS295P1
Purified Ab with BSA and Azide 500μL
Each for $567.50
Add to cart
 
MS295R7
View Documents
Thermo Scientific™
MS295R7
Ready-to-use for IHC (UVLP) 7mL
Each for $425.50
Add to cart
 
MS295R1
View Documents
Thermo Scientific™
MS295R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
MS295P1ABX
View Documents
Thermo Scientific™
MS295P1ABX
Ab without BSA/Azide (Frozen) 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS295PABX
View Documents
Thermo Scientific™
MS295PABX
Ab without BSA/Azide (Frozen) 200μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS295P0
View Documents
Thermo Scientific™
MS295P0
Purified Ab with BSA and Azide 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Provide accurate, reproducible results with the Thermo Scientific™ Troponin T, Cardiac Isoform Ab-1, Mouse Monoclonal Antibody.

Troponin T is a thin filament regulatory protein of striated muscle that binds to troponin C and I and to tropomyosin and is required for calcium-dependent ATPase activity of myofibrillar proteins. It is important in proper muscle function and myofibril formation. In the human heart, four cTnT isoforms are expressed in a developmentally regulated manner. This expression in the human heart is affected by heart failure.

Host Species: Mouse

Clone: 13-11

Isotype: IgG1

Species Reactivity: Human, Pig, Dog, Rabbit, Rat, Mouse, Guinea Pig, Chicken, Sparrow and Goldfish. Does not react with frog.

Epitope: FMPNLVPPKI (5' region of the cardiac gene exon 10)

Immunogen: Purified rabbit cardiac troponin T isoform (TnT4R)

Positive Control: Cardiac muscle

Cellular Localization: Mostly over the I bands in myofibrils Ab-1 specifically recognizes the cardiac isoform of Troponin T (cTnT).

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Electron Microscopy
Specifications

Specifications

Troponin T, Cardiac Isoform Ab-1
Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
IVD
Primary
IgG1
13-11
Monoclonal
Murine
Muscle Biology
Purified rabbit cardiac troponin T isoform (TnT4R)
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit