Learn More
Troponin T type 1 (slow skeletal) Mouse anti-Human, Alexa Fluor™ 532, Clone: 2G3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP229729AF532
Catalog No. NP229729A32
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Troponin T type 1 (slow skeletal) Monoclonal antibody specifically detects Troponin T type 1 (slow skeletal) in Human samples. It is validated for Western Blot,ELISA.
Specifications
Specifications
Troponin T type 1 (slow skeletal)
2G3
Western Blot, ELISA
ANMtroponin T, slow skeletal muscle, FLJ98147, MGC104241, Slow skeletal muscle troponin T, STNT, TNTS, TNTtroponin T1, skeletal, slow, troponin T type 1 (skeletal, slow), troponin-T1, skeletal, slow
Mouse
IgG2b
Store at 4°C in the dark.
Monoclonal
Human cardiac Troponin T No cross reactivity with human skeletal Troponin T.
ELISA, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
TNNT1
Human cardiac Troponin T.
0.1 mL
Primary
7138
Human