Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troponin T type 1 (slow skeletal) Mouse anti-Human, Alexa Fluor™ 594, Clone: 2G3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP229729AF594

Catalog No. NP229729A94


Add to cart

Description

Description

Troponin T type 1 (slow skeletal) Monoclonal antibody specifically detects Troponin T type 1 (slow skeletal) in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Troponin T type 1 (slow skeletal)
2G3
Western Blot, ELISA
Conjugated, Purified
TNNT1
Human cardiac Troponin T.
Protein A or G purified
Cytoskeleton Markers, Stem Cell Markers
Primary
7138.0
Human
ELISA, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
ANMtroponin T, slow skeletal muscle, FLJ98147, MGC104241, Slow skeletal muscle troponin T, STNT, TNTS, TNTtroponin T1, skeletal, slow, troponin T type 1 (skeletal, slow), troponin-T1, skeletal, slow
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human cardiac Troponin T No cross reactivity with human skeletal Troponin T.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit