Learn More
TRPC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific OST00066G
Catalog No. PIOST00066G
Add to Cart
Description
Description
Reconstitute with 500 μL of distilled water.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TRPC1
Polyclonal
Unconjugated
Trpc1, Transient receptor potential protein
Rabbit
Ammonium sulfate precipitation
RUO
399136
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Immunohistochemistry, Western Blot
1 mg/mL
trpc1.L
trpc1.L
A synthetic peptide from c-terminal region of xenopus TRP-1
500 μg
Primary
X. laevis
Antibody
IgG
Recommendations
Recommendations