Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRPC4 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | TRPC4 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB024642
|
Novus Biologicals
NBP295240100UL |
100 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB024643
|
Novus Biologicals
NBP29524020UL |
20 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
TRPC4 Polyclonal antibody specifically detects TRPC4 in Human, Mouse samples. It is validated for Western BlotSpecifications
TRPC4 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
hTrp4, hTrp-4, MGC119570, MGC119571, MGC119572, MGC119573, short transient receptor potential channel 4, transient receptor potential 4, transient receptor potential cation channel, subfamily C, member 4, TRP4HTRP4, TrpC4, Trp-related protein 4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 768-977 of human TRPC4 (NP_057263.1). | |
Primary | |
Store at -20°:C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
PBS (pH 7.3), 50% glycerol | |
7223 | |
IgG | |
Affinity purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title