Learn More
TRPM1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific OSR00186W
Catalog No. PIOSR00186W
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
Reconstitute with 100 μL of distilled water. Centrifuge to remove any insoluble material.
The protein encoded by this gene is similar to the transient receptor potential (TRP) calcium channel family members. The expression of this protein is inversely correlated with melanoma aggressiveness, suggesting that it suppresses melanoma metastasis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TRPM1
Polyclonal
Trpm1
Transient receptor potential cation channel, subfamily M, member 1 variant delta1066-1071, Trpm1, LTRPC1, MLSN1, transient receptor potential cation c
Rabbit
100 μL
Primary
Mouse, Rat
Antibody
IgG
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
Q2TV84, Q2WEA5
TRPM1
A synthetic peptide from aa region 750-800 of rat TRPM1 conjugated to an immunogenic carrier protein was used as the antigen
RUO
17364, 361586
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions
Product Suggestions