Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRPM4 Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Supplier: Thermo Scientific OST00027W
Description
Glycerol (1:1) may be added for additional stability. OST00027W can be used with blocking peptide GST00013P.
Specifications
TRPM4 | |
Polyclonal | |
Trpm4 | |
Melastatin-like 2 | |
Sheep | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q7TN37, Q9ESQ5 | |
TRPM4 | |
A synthetic peptide from mouse transient receptor potential cation channel subfamily M member 4 | |
RUO | |
171143, 68667 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title