Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRPM8 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | TRPM8 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB022835
|
Novus Biologicals
NBP294139100UL |
100 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB022836
|
Novus Biologicals
NBP29413920UL |
20 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
TRPM8 Polyclonal antibody specifically detects TRPM8 in Human samples. It is validated for Western BlotSpecifications
TRPM8 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
Long transient receptor potential channel 6, LTrpC-6, LTRPC6, MGC2849, short form of the TRPM8 cationic channel, transient receptor potential cation channel, subfamily M, member 8, Transient receptor potential p8, transient receptor potential subfamily M member 8, trp-p8, TRPP8transient receptor potential cation channel subfamily M member 8 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 980-1104 of human TRPM8 (NP_076985.4). | |
Primary | |
Store at -20°:C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Tumor Suppressors | |
PBS (pH 7.3), 50% glycerol | |
79054 | |
IgG | |
Affinity purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title