Learn More
TRPS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 200031AP
Catalog No. 50-173-2650
Add to cart
Description
Description
The antibody recognizes the C-term of TRPS1.
This gene encodes a transcription factor that represses GATA-regulated genes and binds to a dynein light chain protein. Binding of the encoded protein to the dynein light chain protein affects binding to GATA consensus sequences and suppresses its transcriptional activity. Defects in this gene are a cause of tricho-rhino-phalangeal syndrome (TRPS) types I-III.
Specifications
Specifications
TRPS1
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q925H1, Q9UHF7
GC79, LGCR, TRPS1, Zinc finger protein GC79
Rabbit
IgG
150 μL
-20°C
Primary
299897, 7227, 83925
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
TRPS1
Liquid
TRPS1
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat