Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRPV4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534292
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Hamster, Panda, Bovine, Dog, Horse, Pig (100%) Elephant, Rabbit (95%).
TRPV4 (Transient Receptor Potential Cation Channel Subfamily V Member 4) is a cation-selective channel, activated in response to systemic osmotic pressure. Non-selective calcium permeant cation channel probably involved in osmotic sensitivity and mechanosensitivity. TRPV4 is a member of the OSM9-like transient receptor potential channel (OTRPC) subfamily in the transient receptor potential (TRP) superfamily of ion channels. The TRPV4 protein is a Ca2+-permeable, nonselective cation channel that is thought to be involved in the regulation of systemic osmotic pressure. Mutations in the TRPV4 gene are the cause of spondylometaphyseal and metatropic dysplasia and hereditary motor and sensory neuropathy type IIC. Diseases associated with TRPV4 include Metatropic Dysplasia and Spondylometaphyseal Dysplasia, Kozlowski Type. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
TRPV4 | |
Polyclonal | |
Unconjugated | |
Trpv4 | |
0610033B08Rik; BCYM3; CMT2C; HMSN2C; LOW QUALITY PROTEIN: transient receptor potential cation channel subfamily V member 4; OSM9-like transient receptor potential channel 4; osm-9-like TRP channel 4; osmosensitive transient receptor potential channel 4; Otrpc4; SMAL; SPSMA; SSQTL1; Transient receptor potential cation channel subfamily V member 4; transient receptor potential cation channel, subfamily 5, member 4; transient receptor potential cation channel, subfamily V, member 4; transient receptor potential protein 12; Trp12; Trpv4; trpv4 channel; Vanilloi; vanilloid receptor-like channel 2; Vanilloid receptor-like protein 2; vanilloid receptor-related osmotically activated channel; vanilloid receptor-related osmotically activated channel (Vroac); vanilloid receptor-related osmotically activated channel protein; vanilloid receptor-related osmotically-activated channel; Vrl2; VRL-2; VROAC; VR-OAC; wu:fp52e02 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100066552, 100126845, 100751070, 486308, 540259, 59341, 63873, 66026 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.9 mg/mL | |
PBS with 0.1% sodium azide | |
Q9EPK8, Q9ERZ8, Q9HBA0 | |
Trpv4 | |
Synthetic 20 amino acid peptide from internal region of human TRPV4. | |
50 μg | |
Primary | |
Human, Mouse, Rat, Canine, Monkey, Bovine, Hamster, Pig, Horse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction