Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRPV5 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $320.71

Specifications

Antigen TRPV5
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry
Applications Western Blot, Immunohistochemistry
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123438
View Documents
Novus Biologicals
NBP30926825UL
25 μg
Each for $106.19
Add to cart
 
NB123437
View Documents
Novus Biologicals
NBP309268100UL
100 μg
Each for $320.71
Add to cart
 
Description

Description

TRPV5 Polyclonal antibody specifically detects TRPV5 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

TRPV5
Western Blot, Immunohistochemistry
Purified
RUO
PBS buffer, 2% sucrose
56302
Polyclonal
Affinity purified
Western Blot 1.0 ug/ml, Immunohistochemistry
Unconjugated
Rabbit
Human
Calcium transport protein 2, calcium transporter 2, ECaC, ECaC1, ECAC1CaT2, Epithelial calcium channel 1CAT2, Osm-9-like TRP channel 3, OTRPC3, transient receptor potential cation channel subfamily V member 5, transient receptor potential cation channel, subfamily V, member 5, TrpV5
The immunogen is a synthetic peptide directed towards the N terminal region of human TRPV5 (NP_062815). Peptide sequence MLQQKRILESPLLRASKENDLSVLRQLLLDCTCDVRQRGALGETALHIAA
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit