Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRRAP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578246
Description
Positive Control: Jurkat, Raji, NCI-H929, 293T, A431, HeLa, HepG2 Predicted Reactivity: Mouse (98%), Rat (98%), Japanese Medaka (89%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TRRAP is an adapter protein, which is found in various multiprotein chromatin complexes with histone acetyltransferase activity (HAT), which gives a specific tag for epigenetic transcription activation. Component of the NuA4 histone acetyltransferase complex which is responsible for acetylation of nucleosomal histones H4 and H2A. TRRAP plays a central role in MYC transcription activation, and also participates in cell transformation by MYC is required for p53/TP53-, E2F1- and E2F4-mediated transcription activation and is also involved in transcription activation mediated by the adenovirus E1A, a viral oncoprotein that deregulates transcription of key genes. TRRAP probably acts by linking transcription factors such as E1A, MYC or E2F1 to HAT complexes such as STAGA thereby allowing transcription activation, however, it is probably not required in the steps following histone acetylation in processes of transcription activation. TRRAP may be required for the mitotic checkpoint and normal cell cycle progression. TRRAP is a component of a SWR1-like complex that specifically mediates the removal of histone H2A.Z/H2AFZ from the nucleosome.
Specifications
TRRAP | |
Polyclonal | |
Unconjugated | |
TRRAP | |
350/400 kDa PCAF-associated factor; AI481500; OTTHUMP00000197294; OTTHUMP00000197295; PAF350/400; PAF400; STAF40; Tra1; Tra1 homolog; transactivation/transformation-domain associated protein; transformation/transcription domain associated protein; transformation/transcription domain-associated protein; TR-AP; TRRAP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8295 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ChIP Assay, Immunoprecipitation, Western Blot | |
1.0 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q9Y4A5 | |
TRRAP | |
Recombinant protein encompassing a sequence within the center region of human TRRAP. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction