Learn More
Aviva Systems Biology Corp TRUB2 Antibody - middle region (ARP48348_T100) | 100uL
Supplier:  Aviva Systems Biology Corp ARP48348_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TRUB2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TRUB2. Alternate names: CLONE24922, RP11-339B21.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59800