Learn More
Vector Laboratories Vector TrueVIEW Autofluorescence Quenching Kit with DAPI
SDP
Supplier:  Vector Laboratories SP8500
Encompass_Preferred
The TrueVIEW Autofluorescence Quenching Kit with DAPI provides a novel way to diminish unwanted autofluorescence from non-lipofuscin sources and dramatically improve signal-to-noise ratio. It removes unwanted autofluorescence in tissue sections due to aldehyde fixation, red blood cells, and structural elements such as collagen and elastin. The quenching action of the kit reagents provides a clear, unambiguous, “true view” localization of the target antigen.
Catalog No. NC1743636