Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Expedeon TruePrime™ Single Cell WGA Kit version 2.0

Uses reliable method to achieve accurate genome amplification from single cells.

Specifications

Product Type TruePrime™ Single Cell WGA Kit version 2.0
Kit Type WGA kit
Polymerase Phi29 DNA polymerase and TthPrimPol
Sample Type Single cell
Storage Requirements -20°C
View More Specs

Products 1
Catalog Number Mfr. No. No. of Reactions Includes Price Quantity & Availability  
Catalog Number Mfr. No. No. of Reactions Includes Price Quantity & Availability  
EX351025 View Documents Expedeon
351025
25 Reaction buffer (1 x 140μL), Buffer L1 (1 x 70μL), DTT (1 x 50μL), Buffer N (1 x 70μL), dNTPs(1 x 140μL), Water (1 x 750μL), Enzyme 1 (1 x 140μL), Enzyme 2 (1 x 20μL), Control 1 (1 x 50μL) and Control 2 (1 x 50μL) This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • Uses multiple displacement amplification method
  • Consists of ten components, including the enzymes and buffers to perform the amplification reactions
  • Uses alkaline incubation to allow cell lysis and DNA denaturation of genomic DNA with very low DNA fragmentation
  • Contains Reaction buffer, Buffer L1, DTT, Buffer N, dNTPs, Water, Enzyme 1, Enzyme 2, Control 1 and Control 2
Specifications

Specifications

TruePrime™ Single Cell WGA Kit version 2.0
Phi29 DNA polymerase and TthPrimPol
-20°C
DNA amplification
WGA kit
Single cell
Multiple displacement amplification method
Safety and Handling

Safety and Handling

ShelfLife : Two years

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit